2-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)nicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)pyridine-3-carboxylic acid, 2-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)-3-carboxypyridine

OR01710-100MG
APOSOR01710-100MGEA 302 EUR
APOSOR01710-100MG
2-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)nicotinic acid
2-({2-[(tert-Butoxycarbonyl)amino]ethyl}amino)nicotinic acid
Formel: C₁₃H₁₉N₃O₄
Molecular Weight: 281,308 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06245545
CAS-Nummer: 904815-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR