1-(2-Nitrophenyl)-2-imidazolidinon

Lieferant: Apollo Scientific
Achtung

OR01735-50MG
APOSOR01735-50MGEA 34.8 EUR
APOSOR01735-50MG
1-(2-Nitrophenyl)-2-imidazolidinon
1-(2-Nitrophenyl)-2-imidazolidinon
Formel: C₉H₉N₃O₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 500890-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR