5-Chloro-6-(piperazin-1-yl)nicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Carboxy-5-chloro-6-(piperazin-1-yl)pyridine, 5-Chloro-6-(piperazin-1-yl)pyridine-3-carboxylic acid, 1-(5-Carboxy-3-chloropyridin-2-yl)piperazine
OR01740-250MG
APOSOR01740-250MGEA
325
EUR
APOSOR01740-250MG
5-Chloro-6-(piperazin-1-yl)nicotinic acid
5-Chloro-6-(piperazin-1-yl)nicotinic acid
Formel:
C₁₀H₁₂ClN₃O₂ Molecular Weight: 241.674 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245471 CAS-Nummer: 889953-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...