2,6-Dihydroxy-3-nitrobenzonitril

Lieferant: Apollo Scientific

OR017404-1G
APOSOR017404-1GEA 125 EUR
APOSOR017404-1G
2,6-Dihydroxy-3-nitrobenzonitril
2,6-Dihydroxy-3-nitrobenzonitril
Formel: C₇H₄N₂O₄
Schmelzpunkt: 238 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 233585-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR