2,3-Dimethylquinoxaline-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR01746-5G
APOSOR01746-5GEA 122 EUR
APOSOR01746-5G
2,3-Dimethylquinoxaline-6-carboxylic acid
2,3-Dimethylquinoxaline-6-carboxylic acid
Formel: C₁₁H₁₀N₂O₂
MDL: MFCD02575516
CAS-Nummer: 17635-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR