4-Bromo-1H-pyrazole-5-carboxaldehyde dimer 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,8-Dibromo-4,9-dihydroxy-4H,9H-dipyrazolo[1,5-a:1',5'-d]pyrazine, 3,8-Dibromo-4H,9H-dipyrazolo[1,5-a:1',5'-d]pyrazine-4,9-diol
OR017542-500MG
APOSOR017542-500MGEA
140
EUR
APOSOR017542-500MG
4-Bromo-1H-pyrazole-5-carboxaldehyde dimer 95+%
4-Bromo-1H-pyrazole-5-carboxaldehyde dimer
Formel:
C₈H₆Br₂N₄O₂ Molecular Weight: 349,967 g/mol Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445454 CAS-Nummer: 1212466-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...