2-Amino-3-cyano-4,5-di(furyl)furan
Lieferant: Apollo Scientific
|
Achtung
|
OR01757-250MG
APOSOR01757-250MGEA
92.8
EUR
APOSOR01757-250MG
2-Amino-3-cyano-4,5-di(furyl)furan
2-Amino-3-cyano-4,5-di(furyl)furan
Formel:
C₁₃H₈N₂O₃ Schmelzpunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00099027 CAS-Nummer: 24386-17-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...