1-(2,4-Dichlorophenyl)ethylamine 98%

Lieferant: Apollo Scientific

Synonyme: 1-Amino-1-(2,4-dichlorophenyl)ethane, 2,4-Dichloro-alpha-methylbenzylamine, 1-(2,4-Dichlorophenyl)ethanamine

OR01762-25G
APOSOR01762-25GEA 72.5 EUR
APOSOR01762-25G
1-(2,4-Dichlorophenyl)ethylamine 98%
1-(2,4-Dichlorophenyl)ethylamine
Formel: C₈H₉Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD05215236
CAS-Nummer: 89981-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR