2-(Pyridin-4-yl)-1H-benzimidazole-6-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 2-(Pyridin-4-yl)-1H-benzo[d]imidazole-6-carboxylic acid, 6-Carboxy-2-(pyridin-4-yl)-1H-benzimidazole, 2-(Pyridin-4-yl)-3H-benzimidazole-5-carboxylic acid

OR01765-1G
APOSOR01765-1GEA 119 EUR
APOSOR01765-1G
2-(Pyridin-4-yl)-1H-benzimidazole-6-carboxylic acid
2-(Pyridin-4-yl)-1H-benzimidazole-6-carboxylic acid
Formel: C₁₃H₉N₃O₂
Molecular Weight: 239.229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06245477
CAS-Nummer: 316833-32-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR