6-(Phenylamino)nicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Anilinopyridine-5-carboxylic acid

OR01774-250MG
APOSOR01774-250MGEA 111 EUR
APOSOR01774-250MG
6-(Phenylamino)nicotinic acid
6-(Phenylamino)nicotinic acid
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06245489
CAS-Nummer: 13426-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR