2-(2-Nitrophenyl)-1H-benzimidazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Carboxy-2-(2-nitrophenyl)-1H-benzimidazole

OR01777-500MG
APOSOR01777-500MGEA 203 EUR
APOSOR01777-500MG
2-(2-Nitrophenyl)-1H-benzimidazole-5-carboxylic acid
2-(2-Nitrophenyl)-1H-benzimidazole-5-carboxylic acid
Formel: C₁₄H₉N₃O₄
Molecular Weight: 283,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01159447
CAS-Nummer: 190121-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR