2-(4-Methylsulphonylphenyl)ethylamine hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

OR0178-1G
APOSOR0178-1GEA 116 EUR
APOSOR0178-1G
2-(4-Methylsulphonylphenyl)ethylamine hydrochloride 95+%
2-(4-Methylsulphonylphenyl)ethylamine hydrochloride
Formel: C₉H₁₄ClNO₂S
Molecular Weight: 235.73 g/mol
Schmelzpunkt: 202…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06245499
CAS-Nummer: 849020-96-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR