2-(2-Hydroxyphenyl)-1H-benzimidazole-6-carboxylic acid
Lieferant: Apollo Scientific
Synonyme:
2-(6-Carboxy-1H-benzimidazol-2-yl)phenol, 6-Carboxy-2-(2-hydroxyphenyl)-1H-benzimidazole, 2-(2-Hydroxyphenyl)-3H-benzimidazole-5-carboxylic acid
OR01780-1G
APOSOR01780-1GEA
806
EUR
APOSOR01780-1G
2-(2-Hydroxyphenyl)-1H-benzimidazole-6-carboxylic acid
2-(2-Hydroxyphenyl)-1H-benzimidazole-6-carboxylic acid
Formel:
C₁₄H₁₀N₂O₃ Molecular Weight: 254,241 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660385 CAS-Nummer: 158040-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...