3-Bromo-2-phenylimidazo[1,2-b]pyridazine

Lieferant: Apollo Scientific
Achtung

OR01787-1G OR01787-500MG
APOSOR01787-1GEA 393 EUR
APOSOR01787-1G APOSOR01787-500MG
3-Bromo-2-phenylimidazo[1,2-b]pyridazine
3-Bromo-2-phenylimidazo[1,2-b]pyridazine
Formel: C₁₂H₈BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09832525
CAS-Nummer: 802580-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR