4-Amino-3-chloro-5-methylbenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR017872-250MG
APOSOR017872-250MGEA 21.9 EUR
APOSOR017872-250MG
4-Amino-3-chloro-5-methylbenzoic acid 95%
4-Amino-3-chloro-5-methylbenzoic acid
Formel: C₈H₈ClNO₂
Schmelzpunkt: 282…284 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01313709
CAS-Nummer: 157069-52-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR