2-Chloro-6-phenylnicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR01798-1G
APOSOR01798-1GEA 21.9 EUR
APOSOR01798-1G
2-Chloro-6-phenylnicotinonitrile
2-Chloro-6-phenylnicotinonitrile
Formel: C₁₂H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01316320
CAS-Nummer: 43083-14-3
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR