Ethyl-7-octenoate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR01812-1G
APOSOR01812-1GEA
0
EUR
APOSOR01812-1G
Ethyl-7-octenoate 97%
Ethyl-7-octenoate
Formel:
C₁₀H₁₈O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00671846 CAS-Nummer: 35194-38-8 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...