6-Bromnicotinsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Brom-3-pyridincarbonsäure, 6-Brompyridin-3-carbonsäure
OR0182-25G
OR0182-5G
OR0182-100G
APOSOR0182-25GEA
40.6
EUR
APOSOR0182-25G
APOSOR0182-5G
APOSOR0182-100G
6-Bromnicotinsäure
6-Bromnicotinsäure
Formel:
C₆H₄BrNO₂ Schmelzpunkt: 179…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01927100 CAS-Nummer: 6311-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...