3-Chlor-4-hydroxybenzonitril

Lieferant: Apollo Scientific
Achtung

OR018202-1G
APOSOR018202-1GEA 21.9 EUR
APOSOR018202-1G
3-Chlor-4-hydroxybenzonitril
3-Chlor-4-hydroxybenzonitril
Formel: C₇H₄ClNO
Molecular Weight: 153,57 g/mol
Schmelzpunkt: 150…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01567246
CAS-Nummer: 2315-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR