2-Methylnon-1-ene

Lieferant: Apollo Scientific

OR01853-1G
APOSOR01853-1GEA 661 EUR
APOSOR01853-1G
2-Methylnon-1-ene
2-Methylnon-1-ene
Formel: C₁₀H₂₀
Molecular Weight: 140,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00027320
CAS-Nummer: 2980-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR