3-(3-Bromophenyl)-1-propene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Allyl-3-bromobenzene

OR01860-100MG
APOSOR01860-100MGEA 39.3 EUR
APOSOR01860-100MG
3-(3-Bromophenyl)-1-propene
3-(3-Bromophenyl)-1-propene
Formel: C₉H₉Br
Molecular Weight: 197,08 g/mol
Siedepunkt: 45…47 °C/0,05mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01319540
CAS-Nummer: 18257-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR