2-[4-(1,3-Dithiolan-2-yl)phenoxy]-N'-hydroxyethanimidamide

Lieferant: Apollo Scientific
Achtung

Synonyme: [4-(1,3-Dithiolan-2-yl)phenoxy]acetamidoxime, 2-[4-(1,3-Dithiolan-2-yl)phenoxy]-N'-hydroxyacetamidine, 2-{4-[2-Amino-2-(hydroxyimino)ethoxy]phenyl}-1,3-dithiolane

OR018627-1G
APOSOR018627-1GEA 63.8 EUR
APOSOR018627-1G
2-[4-(1,3-Dithiolan-2-yl)phenoxy]-N'-hydroxyethanimidamide
2-[4-(1,3-Dithiolan-2-yl)phenoxy]-N'-hydroxyethanimidamide
Formel: C₁₁H₁₄N₂O₂S₂
Molecular Weight: 270,371 g/mol
Schmelzpunkt: 113…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00663347
CAS-Nummer: 262607-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR