3-(4-Bromophenyl)-1-propene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Allyl-4-bromobenzene
OR01868-1G
APOSOR01868-1GEA
580
EUR
APOSOR01868-1G
3-(4-Bromophenyl)-1-propene
3-(4-Bromophenyl)-1-propene
Formel:
C₉H₉Br Molecular Weight: 197,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319542 CAS-Nummer: 2294-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...