4-(4-Bromophenyl)-1-butene
Lieferant: Apollo Scientific
Synonyme:
1-Bromo-4-(but-3-en-1-yl)benzene
OR01869-1G
APOSOR01869-1GEA
763
EUR
APOSOR01869-1G
4-(4-Bromophenyl)-1-butene
4-(4-Bromophenyl)-1-butene
Formel:
C₁₀H₁₁Br Molecular Weight: 211,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319543 CAS-Nummer: 15451-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...