N-(4-Bromobut-1-yl)-4-nitrophthalimide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Bromobut-1-yl)-5-nitro-1H-isoindole-1,3(2H)-dione, 2-(4-Bromobut-1-yl)-5-nitroisoindolin-1,3-dione

OR018728-1G
APOSOR018728-1GEA 92.8 EUR
APOSOR018728-1G
N-(4-Bromobut-1-yl)-4-nitrophthalimide
N-(4-Bromobut-1-yl)-4-nitrophthalimide
Formel: C₁₂H₁₁BrN₂O₄
Molecular Weight: 327,131 g/mol
Schmelzpunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01313849
CAS-Nummer: 125207-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR