4-(3-Chlorophenyl)-1-butene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(But-3-en-1-yl)-3-chlorobenzene

OR01885-250MG
APOSOR01885-250MGEA 71.2 EUR
APOSOR01885-250MG
4-(3-Chlorophenyl)-1-butene
4-(3-Chlorophenyl)-1-butene
Formel: C₁₀H₁₁Cl
Molecular Weight: 166,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01319546
CAS-Nummer: 91426-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR