N-Boc-4-brom-2-methylanilin
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl (4-bromo-2-methylphenyl)carbamate, tert-Butyl 4-bromo-2-methylphenylcarbamate, 4-Bromo-2-methylaniline, N-BOC protected
OR018874-25G
OR018874-5G
APOSOR018874-25GEA
179
EUR
APOSOR018874-25G
APOSOR018874-5G
N-Boc-4-brom-2-methylanilin
N-Boc-4-brom-2-methylanilin
Formel:
C₁₂H₁₆BrNO₂ Molecular Weight: 286.1700 g/mol Schmelzpunkt: 159…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01765429 CAS-Nummer: 306937-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...