2-Acetyl-4-nitroindane-1,3-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Acetyl-4-nitro-1H-indene-1,3(2H)-dione, 2-Acetyl-2,3-dihydro-4-nitro-1,3-dioxo-1H-indene

OR018931-250MG
APOSOR018931-250MGEA 21.9 EUR
APOSOR018931-250MG
2-Acetyl-4-nitroindane-1,3-dione
2-Acetyl-4-nitroindane-1,3-dione
Formel: C₁₁H₇NO₅
Molecular Weight: 233,177 g/mol
Schmelzpunkt: 145…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00728684
CAS-Nummer: 25125-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR