4-(4-Chlorophenyl)-1-butene

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(But-3-en-1-yl)chlorobenzene

OR01895-1G
APOSOR01895-1GEA 679 EUR
APOSOR01895-1G
4-(4-Chlorophenyl)-1-butene
4-(4-Chlorophenyl)-1-butene
Formel: C₁₀H₁₁Cl
Molecular Weight: 166,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01319548
CAS-Nummer: 3047-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR