Ethyl-1,2,4-oxadiazole-3-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 1,2,4-oxadiazole-3-carboxylate
OR0191-1G
APOSOR0191-1GEA
144
EUR
APOSOR0191-1G
Ethyl-1,2,4-oxadiazole-3-carboxylate 95%
Ethyl-1,2,4-oxadiazole-3-carboxylate
Formel:
C₅H₆N₂O₃ Molecular Weight: 142.12 g/mol Siedepunkt: 88…90 °C/2.25mmHg Schmelzpunkt: 41…43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245497 CAS-Nummer: 39512-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...