2-Amino-4,5-dimethyl-3-furonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4,5-dimethylfuran-3-carbonitrile, 2-Amino-4,5-dimethyl-3-furancarbonitrile

OR01921-250MG
APOSOR01921-250MGEA 21.9 EUR
APOSOR01921-250MG
2-Amino-4,5-dimethyl-3-furonitrile
2-Amino-4,5-dimethyl-3-furonitrile
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Schmelzpunkt: 163…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00274264
CAS-Nummer: 5117-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR