1-(2,4-Dichlorophenyl)biguanide hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(2,4-Dichlorophenyl)imidodicarbonimidic diamide hydrochloride
OR0193-100G
OR0193-25G
OR0193-5G
APOSOR0193-100GEA
374
EUR
APOSOR0193-100G
APOSOR0193-25G
APOSOR0193-5G
1-(2,4-Dichlorophenyl)biguanide hydrochloride
1-(2,4-Dichlorophenyl)biguanide hydrochloride
Formel:
C₈H₉Cl₂N₅·ClH Schmelzpunkt: 259,5…263.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00203356 CAS-Nummer: 6634-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...