1-(2,4-Dichlorophenyl)biguanide hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(2,4-Dichlorophenyl)imidodicarbonimidic diamide hydrochloride

OR0193-100G OR0193-25G OR0193-5G
APOSOR0193-100GEA 374 EUR
APOSOR0193-100G APOSOR0193-25G APOSOR0193-5G
1-(2,4-Dichlorophenyl)biguanide hydrochloride
1-(2,4-Dichlorophenyl)biguanide hydrochloride
Formel: C₈H₉Cl₂N₅·ClH
Schmelzpunkt: 259,5…263.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00203356
CAS-Nummer: 6634-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR