4-(2-Cyanophenyl)-2-methyl-1-butene

Lieferant: Apollo Scientific
Achtung

OR01943-250MG
APOSOR01943-250MGEA 290 EUR
APOSOR01943-250MG
4-(2-Cyanophenyl)-2-methyl-1-butene
4-(2-Cyanophenyl)-2-methyl-1-butene
Formel: C₁₂H₁₃N
Siedepunkt: 100…115 °C/0,02mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00671927
CAS-Nummer: 731772-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR