5-Chloro-6-(3-chlorophenyl)pyridazin-3(2H)-one 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR0196-100MG
APOSOR0196-100MGEA
46.4
EUR
APOSOR0196-100MG
5-Chloro-6-(3-chlorophenyl)pyridazin-3(2H)-one 95+%
5-Chloro-6-(3-chlorophenyl)pyridazin-3(2H)-one
Formel:
C₁₀H₆Cl₂N₂O Schmelzpunkt: >250 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245503 CAS-Nummer: 886497-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...