Ethyl-5-aminoimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: Apollo Scientific
Achtung

OR01963-1G OR01963-250MG OR01963-5G
APOSOR01963-1GEA 72.5 EUR
APOSOR01963-1G APOSOR01963-250MG APOSOR01963-5G
Ethyl-5-aminoimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-5-aminoimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₀H₁₁N₃O₂
Schmelzpunkt: 216…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09842618
CAS-Nummer: 1000017-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR