1-(2-Methylimidazo[1,2-a]pyridin-3-yl)ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Acetyl-2-methylimidazo[1,2-a]pyridine

OR019646-250MG
APOSOR019646-250MGEA 21.9 EUR
APOSOR019646-250MG
1-(2-Methylimidazo[1,2-a]pyridin-3-yl)ethanone
1-(2-Methylimidazo[1,2-a]pyridin-3-yl)ethanone
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,2000 g/mol
Schmelzpunkt: 112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00665892
CAS-Nummer: 29096-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR