3-Amino-4-hydroxybenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-4-hydroxybenzonitrile, 2-Amino-4-cyanophenol, 2-Amino-4-cyano-phenol
OR01965-1G
OR01965-5G
APOSOR01965-1GEA
52.2
EUR
APOSOR01965-1G
APOSOR01965-5G
3-Amino-4-hydroxybenzonitril
3-Amino-4-hydroxybenzonitril
Formel:
C₇H₆N₂O Schmelzpunkt: 151…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234273 CAS-Nummer: 14543-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...