8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific

OR01966-1G
APOSOR01966-1GEA 342 EUR
APOSOR01966-1G
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₄BrClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09836172
CAS-Nummer: 1000017-98-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR