Ethyl-4,6-dichloro-5-nitronicotinate
Lieferant: Apollo Scientific
|
Achtung
|
OR01969-1G
OR01969-250MG
APOSOR01969-1GEA
119
EUR
APOSOR01969-1G
APOSOR01969-250MG
Ethyl-4,6-dichloro-5-nitronicotinate
Ethyl-4,6-dichloro-5-nitronicotinate
Formel:
C₈H₆Cl₂N₂O₄ Schmelzpunkt: 36…38,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09842620 CAS-Nummer: 154012-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...