Ethyl-3-({[(2-chloroethyl)amino]carbonyl}amino)benzoate
Lieferant: Apollo Scientific
OR01974-1G
APOSOR01974-1GEA
58
EUR
APOSOR01974-1G
Ethyl-3-({[(2-chloroethyl)amino]carbonyl}amino)benzoate
Ethyl-3-({[(2-chloroethyl)amino]carbonyl}amino)benzoate
Formel:
C₁₂H₁₅ClN₂O₃ Molecular Weight: 270,72 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09836176 CAS-Nummer: 803729-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...