3-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR01975-1G
APOSOR01975-1GEA 561 EUR
APOSOR01975-1G
3-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
3-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Formel: C₈H₇ClN₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD09842622
CAS-Nummer: 1000017-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR