N-(1,3-Benzodioxol-5-yl)-N'-(2-chloroethyl)urea

Lieferant: Apollo Scientific

Synonyme: 1-(1,3-Benzodioxol-5-yl)-3-(2-chloroethyl)urea

OR01977-1G
APOSOR01977-1GEA 92.8 EUR
APOSOR01977-1G
N-(1,3-Benzodioxol-5-yl)-N'-(2-chloroethyl)urea
N-(1,3-Benzodioxol-5-yl)-N'-(2-chloroethyl)urea
Formel: C₁₀H₁₁ClN₂O₃
Molecular Weight: 242,659 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09836177
CAS-Nummer: 1000018-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR