4-Nitro-2-propyl-1,3-benzoxazole

Lieferant: Apollo Scientific

OR01984-1G
APOSOR01984-1GEA 183 EUR
APOSOR01984-1G
4-Nitro-2-propyl-1,3-benzoxazole
4-Nitro-2-propyl-1,3-benzoxazole
Formel: C₁₀H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09864801
CAS-Nummer: 1000018-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR