5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-bromobenzoic acid, N-BOC protected
OR019848-5G
APOSOR019848-5GEA
157
EUR
APOSOR019848-5G
5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid 97%
5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid
Formel:
C₁₂H₁₄BrNO₄ Schmelzpunkt: 180…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01619006 CAS-Nummer: 306937-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...