2-Ethyl-4-nitro-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

OR01985-1G
APOSOR01985-1GEA 74.1 EUR
APOSOR01985-1G
2-Ethyl-4-nitro-1,3-benzoxazole
2-Ethyl-4-nitro-1,3-benzoxazole
Formel: C₉H₈N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09864802
CAS-Nummer: 477603-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR