4-Iodo-1-methyl-1H-indazol-3-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-4-iodo-1-methyl-1H-indazole

OR01987-1G
APOSOR01987-1GEA 364 EUR
APOSOR01987-1G
4-Iodo-1-methyl-1H-indazol-3-amine
4-Iodo-1-methyl-1H-indazol-3-amine
Formel: C₈H₈IN₃
Molecular Weight: 273,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864803
CAS-Nummer: 796967-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR