3-Amino-5-brom-1-methyl-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR01988-1G
APOSOR01988-1GEA
106
EUR
APOSOR01988-1G
3-Amino-5-brom-1-methyl-1H-indazol
3-Amino-5-bromo-1-methyl-1H-indazole
Formel:
C₈H₈BrN₃ Schmelzpunkt: 133…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864804 CAS-Nummer: 1000018-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...