5-(Chlorosulphonyl)-2-methylbenzenecarboxylic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(Chlorosulphonyl)-2-methylbenzoic acid

OR01995-1G
APOSOR01995-1GEA 21.9 EUR
APOSOR01995-1G
5-(Chlorosulphonyl)-2-methylbenzenecarboxylic acid
5-(Chlorosulphonyl)-2-methylbenzenecarboxylic acid
Formel: C₈H₇ClO₄S
Molecular Weight: 234,66 g/mol
Schmelzpunkt: 152…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03989642
CAS-Nummer: 89001-57-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR