4-Hydroxy-5-methyl-6-phenylthieno[2,3-d]pyrimidine

Lieferant: Apollo Scientific

OR019951-1G
APOSOR019951-1GEA 125 EUR
APOSOR019951-1G
4-Hydroxy-5-methyl-6-phenylthieno[2,3-d]pyrimidine
4-Hydroxy-5-methyl-6-phenylthieno[2,3-d]pyrimidine
Formel: C₁₃H₁₀N₂OS
Molecular Weight: 242,296 g/mol
Schmelzpunkt: 247…249 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01935060
CAS-Nummer: 306934-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR